missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Neuroplastin Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF7818
This item is not returnable.
View return policy
Description
Neuroplastin Polyclonal specifically detects Neuroplastin in Mouse samples. It is validated for Western Blot.
Specifications
| Neuroplastin | |
| Polyclonal | |
| Unconjugated | |
| P97300 | |
| NPTN | |
| Mouse myeloma cell line NS0-derived recombinant mouse Neuroplastin Gln29-Pro338, predicted Accession # P97300 | |
| 100 μg | |
| Primary | |
| Detects mouse Neuroplastin in direct ELISAs and Western blots. In direct ELISAs, approximately 100% cross-reactivity with recombinant human Neuroplastin-65 is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.05 ug/mL | |
| AW554172, GP55, GP55Stromal cell-derived receptor 1, GP65SDR-1, GP66, MGC102805, neuroplastin, np55, np65, NPTN, SDFR1, SDFR1DKFZp686L2477, SDR1, SDR1stromal cell derived factor receptor 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 27020 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction