missing translation for 'onlineSavingsMsg'
Learn More

Mouse monoclonal [4E3] Anti-Human IgG1 hinge heavy chain (Alkaline Phosphatase), Abcam

Product Code. 32116592
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116592 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116592 Supplier Abcam Supplier No. ab99773500uL
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal [4E3] Anti-Human IgG1 hinge heavy chain (Alkaline Phosphatase)

The product is subject to the following: Abcam Restricted Use Statement

.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen IGHG1
Applications ELISA, Flow Cytometry
Cellular Localization Isoform 1: Secreted; Isoform 2: Cell membrane; Single-pass membrane protein
Classification Monoclonal
Clone '4E3
Conjugate Alkaline Phosphatase (AP)
Formulation pH: 8, Preservative: 0.1% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 0.605% Tris, 0.0095% Magnesium chloride
Gene IGHG1
Gene Accession No. P01857
Host Species Mouse
Immunogen IGHG1
Purification Method Affinity purification
Quantity 500 μL
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Secondary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG1
Shipping Condition Blue Ice
Target Gene IGHG1
Target Molecule IGHG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.