missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Leptin R Antibody, R&D Systems™

Rat Monoclonal Antibody
209.00€ - 532.00€
Specifications
| Antigen | Leptin R |
|---|---|
| Clone | 55305 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18718749
![]() |
R&D Systems
MAB497-SP |
25 μg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18140731
![]() |
R&D Systems
MAB497 |
500 μg |
532.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Leptin R Monoclonal specifically detects Leptin R in Mouse samples. It is validated for Flow Cytometry, ELISA Capture (Matched Antibody Pair).Specifications
| Leptin R | |
| Flow Cytometry 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295 | |
| LEPR | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 55305 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P48356 | |
| 3953 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Leptin R Leu21-Leu840 (predicted) Accession # P48356 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Leptin R in ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title