missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Kininogen Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2206-SP
162.35 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Kininogen Monoclonal specifically detects Kininogen in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Kininogen | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| O08677 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3827 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Immunoprecipitation | |
| 288422 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Kng1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Kininogen Glu21-Ser661 Accession # O08677 | |
| 25 μg | |
| Primary | |
| Detects mouse Kininogen in direct ELISAs and Western blots. In Western blots, reacts with the full length mouse Kininogen (aa 21-661), but not with the light chain alone. In Western blots, no cross-reactivity with recombinant human (rh) Cystatin D, F, SA, SN, recombinant mouse (rm) Cystatin A, B, C, E/M, rmCystatin/Stefin Homolog, rmFetuin A, rhFetuin B, rmHPRG, rhKininogen, or rhKininostatin is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction