missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Integrin beta 3/CD61 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB41182G-100UG
This item is not returnable.
View return policy
Description
Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Integrin beta 3/CD61 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_034705 (Integrin alpha 2b) & O54890 (Integrin beta 3) | |
| ITGB3 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 2b and Integrin beta 3 linked heterodimer Leu32-Arg988 (Integrin alpha 2b) & Glu26-Asp717 (Integrin beta 3) Accession # NP_034705 (Integrin alpha 2b) & O54890 (Integrin beta 3) | |
| RUO | |
| 3690 | |
| Mouse | |
| IgG1 |
| Flow Cytometry | |
| 909114 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD61, CD61 antigen, GP3A, GP3Aplatelet glycoprotein IIIa, GPIIIA, GPIIIaintegrin beta-3, INGRB3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, ITGB3, Platelet membrane glycoprotein IIIa | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Integrin beta 3/CD61 in direct ELISA. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction