missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Mouse Il6 (P08505, 25 a.a. - 211 a.a.) Partial Recombinant Protein

Product Code. 16252120
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 16252120

Brand: Abnova™ P4364.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Sequence: FPTSQVRRGDFTEDTTPNRPVYTTSQVGGLITHVLWEIVEMRKELCNGNSDCMNNDDALAENNLKLPEIQRNDGCYQTGYNQEICLLKISSGLLEYHSYLEYMKNNLKDNKKDKARVLQRDTETLIHIFNQEVKDLHKIVLPTPISNALLTDKLESQKEWLRTKTIQFILKSLEEFLKVTLRSTRQT

Specifications

Accession Number P08505
For Use With (Application) Functional Study, SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 16193
Molecular Weight (g/mol) 22kDa
Name Il6 (Mouse) Recombinant Protein
Preparation Method Escherichia coli expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 10 μg
Immunogen FPTSQVRRGDFTEDTTPNRPVYTTSQVGGLITHVLWEIVEMRKELCNGNSDCMNNDDALAENNLKLPEIQRNDGCYQTGYNQEICLLKISSGLLEYHSYLEYMKNNLKDNKKDKARVLQRDTETLIHIFNQEVKDLHKIVLPTPISNALLTDKLESQKEWLRTKTIQFILKSLEEFLKVTLRSTRQT
Storage Requirements Store at -20°C on dry atmosphere for 2 years. After reconstitution with deionized water, store at 4°C for 1 month or store at -20°C for 6 months. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1EU/μg) (determined by LAL gel clot method)
Gene Alias Il-6
Common Name Il6
Gene Symbol Il6
Biological Activity The ED50 determined by the dose-dependent proliferation of murine 7TD1 cells, was found to be < 0.01ng/mL cells was found to be in the range of 1 x 108 U/mg.
Species E. coli
Recombinant Recombinant
Protein Tag None
Expression System Escherichia coli expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.