Learn More
ACROBiosystems Mouse IL-6 Protein, His Tag
Brand: ACROBiosystems IL6-M5245-20ug
Additional Details : Weight : 0.00010kg
Beschreibung
Interleukin 6 (IL-6) is also known as HGF, BSF2,HSF, IFNB2 and IL-6, originally identified as a B cell differentiation factor, is a multifunctional cytokine that regulates immune responses, hematopoiesis, acute phase responses, and inflammatory reactions.It is secreted by T cells, macrophages , monocytes, fibroblasts,endothelial cells,et.al. to stimulate immune response to trauma, especially burns or other tissue damage leading to inflammation. Interleukin 6 has been shown to interact with interleukin-6 receptor and glycoprotein. IL-6 is relevant to many disease processes such as diabetes,atherosclerosis, depression,Alzheimer's Disease,systemic,lupus erythematosus,prostate cancer and rheumatoid arthritis. Advanced/metastatic cancer patients have higher levels of IL-6 in their blood.Hence there is an interest in developing anti-IL-6 agents as therapy against many of these diseases.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 23.6 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_112445.1 | |
20 μg | |
Phe 25 - Thr 211 | |
IL6,Interleukin-6,BSF2,HSF,IFNB2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |
23.6 kDa | |
Mouse IL-6, His Tag (IL6-M5245) is expressed from human 293 cells (HEK293). It contains AA Phe 25 - Thr 211 (Accession # P08505-1). | |
-20°C | |
IL-6 | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Cytokines&Growth; Factors |