missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Interleukin 6 Recombinant Protein, Invitrogen™
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
Description
Mouse Interleukin 6 Recombinant Protein, SMIL6, from Invitrogen.
Host Species: Baculovirus
Formulation: 1 Vial purified protein (5 ng/ml) with 4% BSA and no preservative
The International reference standard is not available. Calibrated to a Thermo reference standard. Lyophilized product is shipped on cold packs. Stable until expiration if stored desiccated at 2-8°C. Reconstituted cytokine standards should be stored in aliquots at -70°C. Each aliquot should be used only once and discarded afterwards. Stability of reconstituted and frozen cytokine standards may vary. Controls should be run with each standard curve. Sufficient For: ELISA standard curve with replicates or as a control in Western blots. 5000 pg/ml when reconstituted at the recommend volume Reconstitute with distilled water at volume indicated on label. Use or freeze product within 1 hour of reconstitution.
Specifications
Specifications
| For Use With (Application) | Control, ELISA, Western Blot |
| Formulation | 1 Vial purified protein (5 ng/ml) with 4% BSA and no preservative |
| Quantity | 1 Vial |
| Conjugate | Unconjugated |
| Species | Baculovirus |
| Form | Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction