missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-33 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC3626R-100UG
This item is not returnable.
View return policy
Description
IL-33 Monoclonal specifically detects IL-33 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-33 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.2 ug/10^6 cells | |
| Q8BVZ5 | |
| IL33 | |
| E. coli-derived recombinant mouse IL-33 Ser109-Ile266 Accession # Q8BVZ5 | |
| 100 μg | |
| 90865 | |
| Mouse | |
| Supernatant |
| Flow Cytometry | |
| 396118 | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| C9orf26, C9orf26chromosome 9 open reading frame 26 (NF-HEV), DKFZp586H0523, DVS27, DVS27-related protein, IL1F11, IL-1F11, IL33, IL-33, interleukin 33, Interleukin-1 family member 11, interleukin-33, NFHEV, NF-HEV, NF-HEVNFEHEV, Nuclear factor from high endothelial venules, RP11-575C20.2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse IL-33 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction