missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-17/IL-17A Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems IC7211R-100UG
This item is not returnable.
View return policy
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
| IL-17/IL-17A | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| E. coli-derived recombinant mouse IL-17/IL-17A, Thr22-Ala158, Accession # Q62386 | |
| 100 μg | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry (Intracellular Staining) | |
| 881309 | |
| Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
| Q62386 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3605 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction