Learn More
Mouse IgG2B PE-conjugated Antibody, R&D Systems™
Product Code. 18123961
Change view
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18123961 200 Tests 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18123961 Supplier R&D Systems Supplier No. IC0041P
Mouse Monoclonal Antibody has been used in 33 publications
IgG2b Monoclonal specifically detects IgG2b in Mouse samples. It is validated for Control.
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG2b
Classification Monoclonal
Clone 133303
Conjugate R-PE
Dilution Control 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Host Species Mouse
Immunogen KLH
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 200 Tests
Test Specificity Selected for use as mouse IgG 2B isotype control for flow cytometry applications.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2b