Learn More
Mouse IgG2A PE-conjugated Antibody, R&D Systems™
Product Code. 18173701
Change view
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18173701 200 Tests 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18173701 Supplier R&D Systems Supplier No. IC003P
Mouse Monoclonal Antibody has been used in 17 publications
IgG2a Monoclonal specifically detects IgG2a in Mouse samples. It is validated for Control.
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG2a
Classification Monoclonal
Clone 20102
Conjugate R-PE
Dilution Control 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Host Species Mouse
Immunogen KLH
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 200 Tests
Test Specificity Selected for use as mouse IgG 2A isotype control for flow cytometry applications.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG2a