missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG (H+L) Antibody, R&D Systems™

Product Code. 18172861
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18172861 0.5 mg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18172861 Supplier R&D Systems Supplier No. D201CABS2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Donkey Polyclonal Antibody

IgG Polyclonal specifically detects IgG in Mouse samples. It is validated for .
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Hidden
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Donkey
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity This antibody reacts with heavy chains on mouse IgG, and with light chains on all mouse immunoglobulins. No reactivity observed with other mouse serum proteins, or IgG from bovine, chicken, goat, guinea pig, horse, human, rabbit, rat, sheep, or Syrian hamster, by IEP. This antibody is adsorbed against solid phase human IgG, bovine IgG, rabbit IgG, rat IgG, goat IgG, and Syrian hamster IgG.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8degrees C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.