missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Mouse FGF R2 (IIIb) protein, His Tag

Brand:  ACROBiosystems FGB-M52H5-100ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17206210

  • 457.25€ / 100µg
Estimated Shipment: 07-08-2024
to see stock.
Add to basket

Description

Description

Tyrosine-protein kinase that acts as cell-surface receptor for fibroblast growth factors and plays an essential role in the regulation of cell proliferation, differentiation, migration and apoptosis, and in the regulation of embryonic development. Required for normal embryonic patterning, trophoblast function, limb bud development, lung morphogenesis, osteogenesis and skin development. Plays an essential role in the regulation of osteoblast differentiation, proliferation and apoptosis, and is required for normal skeleton development. Promotes cell proliferation in keratinocytes and immature osteoblasts, but promotes apoptosis in differentiated osteoblasts. Phosphorylates PLCG1, FRS2 and PAK4. Ligand binding leads to the activation of several signaling cascades. Activation of PLCG1 leads to the production of the cellular signaling molecules diacylglycerol and inositol 1,4,5-trisphosphate. Phosphorylation of FRS2 triggers recruitment of GRB2, GAB1, PIK3R1 and SOS1, and mediates activation of RAS, MAPK1/ERK2, MAPK3/ERK1 and the MAP kinase signaling pathway, as well as of the AKT1 signaling pathway. FGFR2 signaling is down-regulated by ubiquitination, internalization and degradation. Mutations that lead to constitutive kinase activation or impair normal FGFR2 maturation, internalization and degradation lead to aberrant signaling. Over-expressed FGFR2 promotes activation of STAT1.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.8 kDa. The protein migrates as 40-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

24.8 kDa
Mouse FGFR2 (IIIb), His Tag (FGB-M52H5) is expressed from human 293 cells (HEK293). It contains AA Pro 39 - Leu 243 (Accession # P21803-2).
-20°C
FGF R2 (IIIb)
Mouse
PBS, pH7.4
HEK293
Bio-Markers & CD Antigens
100 μg
Pro 39 - Leu 243
FGF R2 (IIIb),FGFR2B,FGFR2
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers