missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fc gamma RI/CD64 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2074-SP
This item is not returnable.
View return policy
Description
Fc gamma RI/CD64 Polyclonal specifically detects Fc gamma RI/CD64 in Mouse samples. It is validated for Western Blot.
Specifications
| Fc gamma RI/CD64 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD64, CD64 antigen, CD64a, CD64b, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FCGR1B, FcgRI, FcgRIA, FCRI, FcRIA, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2209 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P26151 | |
| Fcgr1 | |
| NS0-derived recombinant mouse Fc gamma RI/CD64 Glu25-Pro297 Accession # P26151 | |
| 25 μg | |
| Primary | |
| Detects mouse Fc gamma RI/CD64 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant human Fc gamma RI is observed and less than 5% cross-reactivity with recombinant mouse (rm) Fc gamma RIIB and rmFc gamma RIII is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction