missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse FABP9/T-FABP Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2750
This item is not returnable.
View return policy
Description
FABP9/T-FABP Monoclonal specifically detects FABP9/T-FABP in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| FABP9/T-FABP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| O08716 | |
| Fabp9 | |
| E. coli-derived recombinant mouse FABP9/T-FABP Met1-Val132 Accession # O08716 | |
| 100 μg | |
| Primary | |
| Detects human FABP9/T-FABP in direct ELISAs and Western blots. In direct ELISAs, 25% cross-reactivity with recombinant human (rh) FABP6 and rhFABP7 is observed and no cross-reactivity with recombinant mouse (rm) FABP4, rmFABP5, recombinant rat (rr) FABP1, rrFABP2, or rhFABP3. | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 324926 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FABP-9, Fatty Acid Binding Protein 9, Testis, Fatty Acid-Binding Protein 9, Lipid-Binding Protein, PERF, PERF 15, PERF15, Testis Lipid Binding Protein, Testis Lipid-Binding Protein, Testis-Type Fatty Acid-Binding Protein, TFABP, T-FABP, TLBP | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 646480 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only