missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse EGF, Recombinant Protein

Product Code. 16439485 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 x 5 μg
5 x 50 μg
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 16439485

Brand: Invitrogen™ A42554

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

E. coli Recombinant Protein

EGF (Epidermal growth factor) exerts its actions by binding to the EGF Receptor, a 170 kDa protein kinase. Activation of EGFR initiates diverse cellular pathways in response to toxic environmental stimuli, or to EGF binding to the receptor, the EGFR forms homo- or heterodimers with other family members. Each dimeric receptor complex initiates a distinct signaling pathway by recruiting different Src homology 2 (SH2) containing effector proteins. EGF is far and wide expressed in kidney, cerebrum, prostrate and salivary glands. EGF acts as a potent mitogenic factor and the phosphorylated receptor recruits adapter proteins like GRB2 that activates complex downstream signaling cascades. EGF activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STAT modules. Research studies suggest the protein may also play important role in activating the NF-kappa-B signaling cascade. Defects in the EGGF gene are the cause of hypomagnesemia type 4 and dysregulation has been associated with the growth and progression of certain cancers.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01133
For Use With (Application) Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot
Formulation protein with 0.02% Tween 80, 140 mM NaCl, 2.7 mM KCl, 5% mannitol, 5% trehalose and no preservative; pH 7.4
Gene ID (Entrez) 1950
Name Mouse EGF
Quantity 5 x 50 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Gene Alias AI790464; beta-urogastrone; Egf; Epidermal growth factor; epidermal growth factor (beta-urogastrone); H-EGF; HOMG4; Pro-epidermal growth factor; Pro-epidermal growth factor precursor (EGF); URG; Urogastrone
Gene Symbol EGF
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.