missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse DC-SIGN/CD209 Antibody, R&D Systems™
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
Specifications
| Antigen | DC-SIGN/CD209 |
| Applications | CyTOF, Flow Cytometry |
| Classification | Monoclonal |
| Clone | MMD3R |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.25μg/106 cells, CyTOF-ready |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. |
| Gene Accession No. | FJ168685 |
| Gene Alias | CD209, CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4L, CLEC4LC-type lectin domain family 4 member L, DCSIGN, DC-SIGN1, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?