missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CXCL2/MIP-2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 16 publications
Brand: R&D Systems AF-452-SP
This item is not returnable.
View return policy
Description
CXCL2/GRO beta/MIP-2/CINC-3 Polyclonal specifically detects CXCL2/GRO beta/MIP-2/CINC-3 in Mouse samples. It is validated for Immunohistochemistry, Neutralization, Immunocytochemistry.
Specifications
| CXCL2/GRO beta/MIP-2/CINC-3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-X-C motif) ligand 2, CINC-2a, CINC3, CINC-3, C-X-C Motif Chemokine 2, GRO beta, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MGSA-beta, MIP2, MIP-2, MIP-2a, MIP2A, MIP2-alpha, SCYB2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2920 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.015-0.075 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P10889 | |
| CXCL2 | |
| E. coli-derived recombinant mouse CXCL2/MIP-2 Ala28-Asn100 Accession # P10889 | |
| 25 μg | |
| Primary | |
| Detects mouse CXCL2/MIP-2 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) GRO alpha , rhGRO beta , rhGRO gamma , recombinant rat (rr) CINC-1, rrCINC-2 alpha , rrCINC-2 beta , and rrCINC-3 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction