missing translation for 'onlineSavingsMsg'
Learn More

Mouse CXCL13/BLC/BCA-1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18709707
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
500 ug
Unit Size:
100µg
1mg
25µg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18709707 100 μg 100µg
18743819 25 μg 25µg
18191741 1 mg 1mg
18140801 500 ug 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18709707 Supplier R&D Systems Supplier No. MAB470R100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CXCL13/BLC/BCA-1 Monoclonal specifically detects CXCL13/BLC/BCA-1 in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL13/BLC/BCA-1
Applications Neutralization, ELISA
Classification Monoclonal
Clone 143614R
Conjugate Unconjugated
Dilution Neutralization 12-30 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O55038
Gene Alias ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1, BCA-1, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLC, BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant)
Gene Symbols Cxcl13
Host Species Rat
Immunogen E. coli-derived recombinant mouse CXCL13 Ile22-Ala109 Accession # O55038
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10563
Test Specificity Detects mouse CXCL13 in ELISAs. In ELISAs, this antibody does not cross-react with recombinant human (rh) CXCL13, rmCXCL11/I-TAC, rmCXCL9/MIG, or rm CXCL12/SDF-1 alpha .
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.