missing translation for 'onlineSavingsMsg'
Learn More

Mouse CD300f/LMIR3 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18769067
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18769067 25 μg 25µg
18091072 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18769067 Supplier R&D Systems Supplier No. AF2788SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

CD300f/LMIR3/CD300LF Polyclonal specifically detects CD300f/LMIR3/CD300LF in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300f/LMIR3/CD300LF
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6SJQ7
Gene Alias CMRF35-like molecule 1, CD300 molecule-like family member f, CD300f, CD300LF, CLIM1, CLM1, CLM-1, DIgR2, IgSF13, IREM1, IREM-1, LMIR3, NKIR, Pigr3
Gene Symbols CD300LF
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD300f/LMIR3 Cys16-Gly188 Accession # Q6SJQ7
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146722
Test Specificity Detects mouse CD300f/LMIR3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) LMIR1, rhLMIR2, rhLMIR3, recombinant mouse LMIR5, and rhLMIR6 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.