missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD300a/LMIR1 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1186V-100UG
This item is not returnable.
View return policy
Description
CD300a/LMIR1 Monoclonal specifically detects CD300a/LMIR1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD300a/LMIR1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| BAC80268 | |
| CD300A | |
| Mouse myeloma cell line NS0-derived recombinant mouse CD300a/LMIR1 Leu28-Arg183 Accession # BAC80268 | |
| RUO | |
| 11314 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 172224 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD300a, CD300a antigenCMRF35H9, CD300a molecule, CLM8, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, IRp60, leukocyte membrane antigen, LMIR1, MAIR-I, Mcipr1, NKRL, Pigr4 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse CD300a/LMIR1 in direct ELISAs. In direct ELISAs, approximately 2% cross-reactivity with recombinant mouse (rm) LMIR2 is observed and no cross-reactivity with recombinant human LMIR1, 2, 3, 4, 5, 6, or rmLMIR3, 4, or 5 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction