missing translation for 'onlineSavingsMsg'
Learn More

Mouse CD2 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  R&D Systems FAB2828G-100UG

312.80 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18736607

  • 378.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD2 Monoclonal specifically detects CD2 in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD2
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P08920
CD2
Mouse myeloma cell line NS0-derived recombinant mouse CD2 Arg23-Ser203 Accession # P08920
RUO
914
Mouse
IgG2a
Flow Cytometry
377830
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
CD2, CD2 antigen, CD2 antigen (p50), sheep red blood cell receptor, CD2 molecule, Erythrocyte receptor, FLJ46032, LFA-2, LFA-3 receptor, lymphocyte-function antigen-2, Rosette receptor, SRBC, T11, T-cell surface antigen CD2, T-cell surface antigen T11/Leu-5
Rat
100 μg
Primary
Detects mouse CD2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CD2 is observed.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only (RUO)