missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCR5 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB18041-100
This item is not returnable.
View return policy
Description
CCR5 Monoclonal specifically detects CCR5 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CCR5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
| C6 rat glioma cell line transfected with mouse CCR5, Accession No. P51682 | |
| 100 μg | |
| 1234.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 225307 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P51682 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse CCR5 transfectants but not the parental cell line in Flow Cytometry and Western blots. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction