missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL24/Eotaxin-2/MPIF-2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB528-100
This item is not returnable.
View return policy
Description
CCL24/Eotaxin-2/MPIF-2 Monoclonal specifically detects CCL24/Eotaxin-2/MPIF-2 in Mouse samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CCL24/Eotaxin-2/MPIF-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-C motif) ligand 24, Ckb-6, Eosinophil chemotactic protein 2, Eotaxin-2, member 24, MPIF2, MPIF-2, MPIF2Myeloid progenitor inhibitory factor 2, MPIF-2Small-inducible cytokine A24, SCYA24CK-beta-6 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6369 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization, ELISA | |
| 106521 | |
| Western Blot 1 ug/mL, Neutralization 0.06-0.3 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9JKC0 | |
| CCL24 | |
| E. coli-derived recombinant mouse CCL24/Eotaxin-2/MPIF-2 Ile24-Val119 Accession # Q9JKC0 | |
| 100 μg | |
| Primary | |
| Detects mouse CCL24/Eotaxin-2/MPIF-2 in Western blots. In Western blots, no cross-reactivity with recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 28, recombinant mouse CCL1, 2, 3, 4, 6, 7, 9/10/MIP-1 gamma , 11, 12, 19, 20, 21, 22, 25, 28, or recombinant rat CCL20 is observed. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction