missing translation for 'onlineSavingsMsg'
Learn More

Mouse Cardiotrophin-1/CT-1 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 3 publications

196.00€ - 524.00€

Specifications

Antigen Cardiotrophin-1/CT-1
Clone 91907
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18788729
GREENER_CHOICE
View Documents
R&D Systems
MAB438-SP
25 μg
196.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18140971
View Documents
R&D Systems
MAB438
500 μg
524.00€
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cardiotrophin-1/CT-1 Monoclonal specifically detects Cardiotrophin-1/CT-1 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

Cardiotrophin-1/CT-1
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
cardiophin 1, cardiotrophin 1, cardiotrophin-1, Cardiotrophin1, CT-1CT1, CTF1
CTF1
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
91907
Western Blot, ELISA
Unconjugated
Rat
Mouse
Q60753
1489
E. coli-derived recombinant mouse Cardiotrophin-1 Ser2-Ala203 Accession # Q60753
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse Cardiotrophin-1 in ELISAs and Western blots. In ELISAs, this antibody shows approximately 20% cross-reactivity with recombinant human (rh) Cardiotrophin-1 and no cross-reactivity with rmIL-11, rmIL-6, rrCNTF, rmLIF, or rmOSM.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.