missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cardiotrophin-1/CT-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF438-SP
This item is not returnable.
View return policy
Description
Cardiotrophin-1/CT-1 Polyclonal specifically detects Cardiotrophin-1/CT-1 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| Cardiotrophin-1/CT-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cardiophin 1, cardiotrophin 1, cardiotrophin-1, Cardiotrophin1, CT-1CT1, CTF1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1489 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.08-0.48 ug/mL | |
| Q60753 | |
| CTF1 | |
| E. coli-derived recombinant mouse Cardiotrophin-1/CT-1 Ser2-Ala203 Accession # Q60753 | |
| 25 μg | |
| Primary | |
| Detects Cardiotrophin-1/CT-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human CT-1 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction