missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Carbonic Anhydrase IX/CA9 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 12 publications
Brand: R&D Systems AF2344-SP
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IX/CA9 Polyclonal specifically detects Carbonic Anhydrase IX/CA9 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
Specifications
| Carbonic Anhydrase IX/CA9 | |
| Polyclonal | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q8VHB5 | |
| Car9 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Carbonic Anhydrase IX/CA9 Gln32-Asp390 Accession # Q8VHB5 | |
| 25 μg | |
| Primary | |
| Detects mouse Carbonic Anhydrase IX/CA9 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human (rh) CA9 is observed, 5% cross-reactivity with recombinant mouse (rm) CA14 is observed, and less than 1% cross-reactivity with rhCA1, -2, -3, -8, -10, -13, rmCA4, and -12 is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CA9, CAIX, CA-IX, Carbonate dehydratase IX, carbonic anhydrase 9, carbonic anhydrase IX, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MN, P54/58N, PMW1, RCC, RCC-associated antigen G250, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 768 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction