missing translation for 'onlineSavingsMsg'
Learn More

Mouse/Canine Integrin β2/CD18 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Product Code. 31841732
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31841732 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31841732 Supplier R&D Systems Supplier No. AF2618R100UG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

Integrin beta 2/CD18 Polyclonal antibody specifically detects Integrin beta 2/CD18 in Canine,Mouse samples. It is validated for Flow Cytometry,Western Blot,Adhesion Activation,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin beta 2/CD18
Applications Flow Cytometry, Western Blot, Adhesion Activation, CyTOF
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation Supplied 0.2mg/mL in 1X PBS with RDF1 and 0.09% Sodium Azide
Gene Alias 959 beta subunit precursor)10, CD18, CD18 antigen, CD18beta 2, cell surface adhesion glycoprotein (LFA-1, CR3, integrin beta-2, integrin, beta 2 (antigen CD18 (p95), lymphocyte function-associated antigen 1;macrophage antigen 1 (mac-1) beta subunit), integrin, beta 2 (complement component 3 receptor 3 and 4 subunit), ITGB2, LAD, LCAMB, leukocyte cell adhesion molecule CD18, leukocyte-associated antigens CD18/11A, CD18/11B, CD18/11C, LFA-1, LFA-1 beta, MAC-1 beta, MF17, MFI7, P150, p150,95 beta
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin Î22, Gln24-Asn702, Accession # P11835
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3689
Target Species Canine, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied
Product Type Antibody
Form Purified
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.