missing translation for 'onlineSavingsMsg'
Learn More

ZNF608, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16162599
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16162599 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16162599 Supplier Abnova Supplier No. H00057507B03P.50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a full-length human ZNF608 protein.

Sequence: MSVNISTAGKGVDPNTVDTYDSGDDWEIGVGNLIIDLDADLEKDRQKFEMNNSTTTTSSSNSKDCGGPASSGAGATAALADGLKFASVQASAPQGNSHKETSKSKVKRSKTSKDANKSLPSAALYGIPEISSTGKRQEVQGRPGEATGMNSALGQSVSSGGSGNPNSNSTSTSTSAATAGAGSCGKSKEEKPGKSQSSRGAKRDKDAGKSRKDKHDLLQGHQNGSGSQAPSGGHLYGFGAKSNGGGASPFHCGGTGSGSVAAAGEVSKSAPDSGLMGNSMLVKKEEEEEESHRRIKKLKTEKVDPLFTVPAPPPPISSSLTPQILPSYFSPSSSNIAAPVEQLLVRTRSVGVNTCEVGVVTEPECLGPCEPGTSVNLEGIVWHETEEGVLVVNVTWRNKTYVGTLLDCTKHDWAPPRFCESPTSDLEMRGGRGRGKRARSAAAAPGSEASFTESRGLQNKNRGGANGKGRRGSLNASGRRTPPNCAAEDIKASPSSTNKRKNKPPMELDLNSSSEDNKPGKRVRTNSRSTPTTPQGKPETTFLDQGCSSPVLIDCPHPNCNKKYKHINGLRYHQAHAHLDPENKLEFEPDSEDKISDCEEGLSNVALECSEPSTSVSAYDQLKAPASPGAGNPPGTPKGKRELMSNGPGSIIGAKAGKNSGKKKGLNNELNNLPVISNMTAALDSCSAADGSLAAEMPKLEAEGLIDKKNLGDKEKGKKANNCKTDKNLSKLKSARPIAPAPAPTPPQLIAIPTATFTTTTTGTIPGLPSLTTTVVQATPKSPPLKPIQPKPTIMGEPITVNPALVSLKDKKKKEKRKLKDKEGKETGSPKMDAKLGKLEDSKGASKDLPGHFLKDHLNKNEGLANGLSESQESRMASIKAEADKVYTFTDNAPSPSIGSASRLECSTLVNGQAPMAPLHVLTQNGAESSAAKTSSPAYSDISDAADDGGSDSRSEGMRSKASSPSDIISSKDSVVKGHSSTTAQSSQLKESHSPYYHSYDPYYSPSYMHPGQVGAPAAGNSGSTQGMKIKKESEEDAEKKDKAEQLDSKKVDHNSASLQPQHQSVITQRHPALAQSLYYGQYAYGLYMDQKSLMATSPAYRQQYEKYYEDQRLAEQKMAQTGRGDCERKSELPLKELGKEETKQKNMPSATISKAPSTPEPNKNHSKLGPSVPNKTEETGKSQLLSNHQQQLQADSFKAKQMENHQLIKEAVEMKSVMDSMKQTGVDPTSRFKQDPDSRTWHHYVYQPKYLDQQKSEELDREKKLKEDSPRKTPNKESGVPSLPVSLTSIKEEPKEAKHPDSQSMEESKLKNDDRKTPVNWKDSRGTRVAVSSPMSQHQSYIQYLHAYPYPQMYDPSHPAYRAVSPVLMHSYPGAYLSPGFHYPVYGKMSGREETEKVNTSPSVNTKTTTESKALDLLQQHANQYRSKSPAPVEKATAEREREAERERDRHSPFGQRHLHTHHHTHVGMGYPLIPGQYDPFQGLTSAALVASQQVAAQASASGMFPGQRRE

Specifications

Antigen ZNF608
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ZNF608 protein.
Formulation PBS with no preservative; pH 7.4
Gene ZNF608
Gene Accession No. NM_020747
Gene Alias DKFZp781C0723/MGC166851/NY-REN-36
Gene Symbols ZNF608
Host Species Mouse
Immunogen ZNF608 (NP_065798.1,1 a.a. ∽ 1512 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57507
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.