missing translation for 'onlineSavingsMsg'
Learn More

STING Mouse anti-Human, PE-CF594, Clone: T3-680, BD

Product Code. 15894229
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15894229 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15894229 Supplier BD Biosciences Supplier No. 565313

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The T3-680 monoclonal antibody specifically recognizes human STING (Stimulator of interferon genes protein) which is also known as Endoplasmic reticulum interferon stimulator (ERIS) and Mediator of IRF3 activation (MITA). STING is encoded by TMEM173 (Transmembrane protein 173). STING is a multi-pass transmembrane adaptor protein which is widely expressed in the endoplasmic reticulum of cells. STING acts as a sensor of cytosolic double-stranded DNA derived from bacteria and viruses and promotes the production of type I interferons, IFN- α and IFN- β . Following activation, STING translocates with TBK1 kinase to perinuclear endosomes. The TBK1 kinase phosphorylates and activates interferon regulatory factors, such as IRF3, and NF-κB, which leads to the induction of type I interferon and other immune response genes that play roles in innate and adaptive immunity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen STING
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone T3-680
Conjugate PE-CF594
Description hSTING; MITA; hMITA; ERIS; MPYS; NET23; TM173; TMEM173
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human STING Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.