Learn More
STAT1, Mouse, Clone: M135, Abnova™
Mouse monoclonal antibody raised against synthetic phosphopeptide of STAT1.
Brand: Abnova MAB1374.100uL
Description
The protein encoded by this gene is a member of the STAT protein family. In response to cytokines and growth factors, STAT family members are phosphorylated by the receptor associated kinases, and then form homo- or heterodimers that translocate to the cell nucleus where they act as transcription activators. This protein can be activated by various ligands including interferon-alpha, interferon-gamma, EGF, PDGF and IL6. This protein mediates the expression of a variety of genes, which is thought to be important for cell viability in response to different cell stimuli and pathogens. Two alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq
Specifications
| STAT1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (1:500) The optimal working dilution should be determined by the end user. | |
| STAT1 | |
| STAT1 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding Y701 of human STAT1. | |
| RUO | |
| 6772 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Western Blot | |
| M135 | |
| Mouse monoclonal antibody raised against synthetic phosphopeptide of STAT1. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| DKFZp686B04100/ISGF-3/STAT91 | |
| Mouse | |
| 100 μL | |
| Primary | |
| This peptide sequence has high homology to the conserved tyrosine site in rat and mouse stat1. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.