missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgG2a, Clone: RG7/1.30, FITC, Secondary Antibody, BD
Description
The Rg7/1.30 antibody reacts specifically with the Fc region of rat IgG2a. It does not react with other Ig isotypes.
Host Species: Mouse
Clone: RG7/1.30
Isotype: IgG2b, κ
Species Reactivity: Rat
Immunogen: Pooled rat IgG
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Rat IgG2a |
| Classification | Monoclonal |
| Clone | RG7/1.30 |
| Concentration | 0.5mg/mL |
| Conjugate | FITC |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?