missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgG1 Heavy Chain Secondary Antibody (MARG1-2), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68397R
This item is not returnable.
View return policy
Description
IgG1 Heavy Chain Monoclonal antibody specifically detects IgG1 Heavy Chain in Rat samples. It is validated for ELISA
Specifications
| IgG1 Heavy Chain | |
| Monoclonal | |
| DyLight 550 | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Rat | |
| Purified |
| ELISA | |
| MARG1-2 | |
| 50mM Sodium Borate | |
| Rat IR27 IgG1 myeloma protein | |
| 0.25 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction