missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rat IgD Secondary Antibody (MARD-3), PerCP, R&D Systems™

Artikelnummer. 30231071 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.25 mL
Pakningsstørrelse:
0.25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
30231071 0.25 mL 0.25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30231071 Leverandør Novus Biologicals Leverandørnr. NBP268388PCP
  • ONLINE EXCLUSIVE PRICE
    748.00€
    706.65€ / 0.25mL
    Save 41.35€
    5% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgD Monoclonal specifically detects IgD in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen IgD
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MARD-3
Conjugate PerCP
Formulation PBS
Gene Alias IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta
Host Species Mouse
Immunogen IR731 myeloma protein
Purification Method Affinity purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Gene ID (Entrez) 3495
Target Species Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.