missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rat Ig κ Light Chain, Clone: MRK-1, PE, Secondary Antibody, BD

Product Code. 15807448
Change view
Click to view available options
Quantity:
0.2 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15807448 0.2 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15807448 Supplier BD Biosciences Supplier No. 553873
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The MRK-1 antibody reacts specifically with rat Igs bearing κ light chain. It does not react withλ light chain or heavy chains. MRK-1 antibody is effective for detection of cell-surface Ig by immunofluorescent staining with flow cytometric analysis. It has not been shown to stimulate B-cell proliferation.

Host Species: Mouse
Clone: MRK-1
Isotype: IgG2a
Species Reactivity: Rat
Immunogen: Pooled rat Ig

ELISA, Flow Cytometry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Rat Ig κ Light Chain
Classification Monoclonal
Clone MRK-1
Concentration 0.2mg/mL
Conjugate PE
Host Species Mouse
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Rat
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.