missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rabbit IgG (H+L) Secondary Antibody, PerCP, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Dot Blot, ELISA, Immunohistochemistry, Western Blot.
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Dot Blot, ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Formulation | PBS |
| Host Species | Mouse |
| Immunogen | Mouse anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed) was produced by repeated immunization with rabbit whole IgG molecule in mouse. |
| Purification Method | Multi-step |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?