missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ptprc, Mouse, Clone: ANK74, Abnova™
Mouse monoclonal antibody raised against native Ptprc.
Brand: Abnova MAB5278.1mL
This item is not returnable.
View return policy
Specifications
| Ptprc | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Ptprc | |
| Ptprc | |
| Native purified Ptprc from IL-2-activated cultured Wag rats NK cells. | |
| RUO | |
| 24699 | |
| Rat | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| ANK74 | |
| Mouse monoclonal antibody raised against native Ptprc. | |
| In culture supernatant (0.05% sodium azide) | |
| CD45/L-CA/Lca/RT7/T200 | |
| Mouse | |
| 1 mL | |
| Primary | |
| This is a rat strain-independent antibody to CD45. This monoclonal antibody recognizes a common epitope of rat CD45, present on all rat leucocytes. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction