missing translation for 'onlineSavingsMsg'
Learn More

PRL, Mouse, Clone: 1/5C4 2D5, Abnova™

Product Code. 16025936
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16025936 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16025936 Supplier Abnova Supplier No. MAB3693.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against recombinant PRL.

Specifications

Antigen PRL
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 1/5C4 2D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant PRL.
Dilution The optimal working dilution should be determined by the end user.
Formulation In BBS
Gene PRL
Gene Symbols PRL
Host Species Mouse
Immunogen Recombinant protein corresponding to human PRL.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5617
Test Specificity This antibody recognizes natural and recombinant human PRL.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.