Learn More
NFKBIA, Mouse, Clone: 39A1413, Abnova™
Mouse monoclonal antibody raised against synthetic phosphopeptide of NFKBIA.
Brand: Abnova MAB0056.100ug
Description
NFKB1 (MIM 164011) or NFKB2 (MIM 164012) is bound to REL (MIM 164910), RELA (MIM 164014), or RELB (MIM 604758) to form the NFKB complex. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA or NFKBIB, MIM 604495), which inactivate NF-kappa-B by trapping it in the cytoplasm. Phosphorylation of serine residues on the I-kappa-B proteins by kinases (IKBKA, MIM 600664, or IKBKB, MIM 603258) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B complex. Activated NFKB complex translocates into the nucleus and binds DNA at kappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or 5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or G purine; and Y is a C or T pyrimidine).[supplied by OMIM
Specifications
| NFKBIA | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| NFKBIA | |
| NFKBIA | |
| Synthetic phosphopeptide corresponding to residues surrounding S32/S36 of human NFKBIA. | |
| RUO | |
| 4792 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunoprecipitation, Western Blot | |
| 39A1413 | |
| Mouse monoclonal antibody raised against synthetic phosphopeptide of NFKBIA. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| IKBA/MAD-3/NFKBI | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.