Learn More
Mouse anti-Mouse IgG2a[b], Clone: 5.7, Secondary Antibody, BD
Description
This antibody reacts specifically with mouse IgG2a of Igh-C [b] haplotype (e.g., C57BL/6, C57BL/10, NOD, SJL). It has been reported not to react with IgG2a of the a, c, d, e, f, g, h, j, n, o, or p haplotypes (e.g., A, AKR, BALB/c, C3H/He, C58, CBA, DBA/1
Host Species: Mouse
Clone: 5.7
Isotype: IgG3, κ
Species Reactivity: Mouse
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Mouse IgG2a[b] |
| Classification | Monoclonal |
| Clone | 5.7 |
| Concentration | 0.5mg/mL |
| Conjugate | Biotin |
| Host Species | Mouse |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |