missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Mouse IgG1[b], Clone: B68-2, Secondary Antibody, BD

Product Code. 15877258
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15877258 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15877258 Supplier BD Biosciences Supplier No. 553533
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The B68-2 antibody reacts specifically with mouse IgG1 of Igh-C[b] haplotype (e.g., C57BL/6, C57BL/10, SJL). It does not react with the Igh-C[a] haplotype (e.g., BALB/c, C58).

Host Species: Mouse
Clone: B68-2
Isotype: IgM, κ
Species Reactivity: Mouse
Immunogen: C57BL/6 Mouse Anti-TNP-KLH Antiserum

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Mouse IgG1[b]
Classification Monoclonal
Clone B68-2
Concentration 0.5mg/mL
Conjugate Biotin
Host Species Mouse
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.