missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Mouse IgD[b], Clone: 217-170, PE, Secondary Antibody, BD

Product Code. 15897248
Change view
Click to view available options
Quantity:
0.2 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15897248 0.2 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15897248 Supplier BD Biosciences Supplier No. 553511
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The 217-170 antibody reacts specifically with mouse IgD of Igh-C[b] haplotype (eg, C57BL/6, C57BL/10, SJL). It does not react with IgD of Igh-C[a] or related haplotypes (eg, BALB/c, AKR, CBA, C3H/He, C58, DBA/1, DBA/2, NZB, PL, RIII), or Igh-C[e] haplotyp

Host Species: Mouse
Clone: 217-170
Isotype: IgG1, κ
Species Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Mouse IgD[b]
Classification Monoclonal
Clone 217-170
Concentration 0.2mg/mL
Conjugate PE
Host Species Mouse
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.