missing translation for 'onlineSavingsMsg'
Learn More

Mos, Mouse, Clone: S3.1, Abnova™

Product Code. 16057076
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16057076 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16057076 Supplier Abnova Supplier No. MAB7045.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against recombinant mos.

Specifications

Antigen Mos
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S3.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant mos.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene mosxe
Gene Alias mos
Gene Symbols mosxe
Host Species Mouse
Immunogen Recombinant protein corresponding to xenopus mos.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 397924
Test Specificity Able to immunoprecipitate MOS from cell/egg extracts. Immunoprecipitation and Western Blot Predicted molecular weight 39 KDa.
Target Species Xenopus species
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.