Learn More
Insulin Mouse, Alexa Fluor 647, Clone: T56-706, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565689
Description
The T56-706 monoclonal antibody specifically binds to insulin, a member of the insulin family of active peptides. Insulin is an evolutionarily conserved peptide hormone that binds to receptors on target cells (primarily adipose and muscle) to promote the absorption of glucose from the blood, thus regulating fat and carbohydrate metabolism. Insulin is produced by β cells in the islets of Langerhans of the pancreas. There, the precursor molecule, preproinsulin, is cleaved to proinsulin that is in turn cleaved to form the mature insulin hormone, which is composed of two peptides (A- and B-chains) linked by 2 disulfide bonds. Mature insulin is stored in granules in the β cells and is released to the blood in response to metabolic signals such as glucose, the amino acids arginine and leucine, and acetylcholine. Catecholamines can regulate blood glucose levels by either stimulating or inhibiting the release of insulin from β cells . The expression of insulin can be used to monitor the pancreatic differentiation of pluripotent stem cells.
Specifications
| Insulin | |
| Monoclonal | |
| 0.2mg/mL | |
| INS; IDDM; ILPR; IRDN; IDDM1; IDDM2; MODY10; Ins2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Mouse, Rat | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunocytochemistry | |
| T56-706 | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Mature human insulin (A- and B-chain) Recombinant Protein | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.