missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leishmania A2, Mouse, Clone: C9, Abnova™
Mouse monoclonal antibody raised against A2.
Brand: Abnova MAB5675.100ug
This item is not returnable.
View return policy
Specifications
| Leishmania A2 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:100) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Protozoa | |
| Lyophilized |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| C9 | |
| Mouse monoclonal antibody raised against A2. | |
| Lyophilized from PBS, pH 7.4 | |
| Native from A2 protein of Leishmania donovani and Leishmania infantum. | |
| RUO | |
| The anti A2 antibody recognizes only the amastigote form of the A2 protein of Leishmania donovani and infantum. There is no cross reactivity with other Leishmania species. This antibody recognizes the family of A2 proteins ranging from 42 to 100 KDa. | |
| Store at 4°C or -20°C on dry atmosphere. After reconstitution with water and glycerol (1:1), store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction