missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clostridium difficile Toxin B, Mouse, Clone: B426M, Abnova™
Mouse monoclonal antibody raised against Clostridium difficile Toxin B.
Brand: Abnova MAB3959.1mg
This item is not returnable.
View return policy
Specifications
| Clostridium difficile Toxin B | |
| Monoclonal | |
| Unconjugated | |
| Recommend dilutions start at 5 ug/mL. The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Clostridium species | |
| Liquid |
| ELISA | |
| B426M | |
| Mouse monoclonal antibody raised against Clostridium difficile Toxin B. | |
| In 10mM PBS, pH 7.2 (0.09% sodium azide) | |
| Clostridium difficile Toxin B. | |
| RUO | |
| Specific for Toxin B of Clostridium difficile. Does not cross-react with Toxin A. | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction