missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Anti-Human IgM, APC, Clone: G20-127, Secondary Antibody, BD
Description
The G20-127 antibody reacts specifically with the heavy chain of human IgM. It does not react with other heavy chain isotypes.
Host Species: Mouse
Clone: G20-127
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
Specifications
| Antigen | Mouse Anti-Human IgM |
| Classification | Monoclonal |
| Clone | G20-127 |
| Conjugate | APC |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Target Species | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?