Learn More
Mouse anti-Human IgM, Clone: G20-127, PE, Secondary Antibody, BD
Beskrivelse
The G20-127 antibody reacts specifically with the heavy chain of human IgM. It does not react with other heavy chain isotypes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G20-127
Isotype: IgG1, κ
Species Reactivity: Human
ELISA, Flow Cytometry
Tekniske data
Tekniske data
| Antigen | Mouse anti-Human IgM |
| Classification | Monoclonal |
| Clone | G20-127 |
| Conjugate | PE |
| Host Species | Mouse |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Target Species | Human |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.