missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG4, Clone: JDC-14, AKP, Secondary Antibody, BD
Description
The JDC-14 antibody reacts specifically with human IgG4. It does not react with other Ig isotypes.
Host Species: Mouse
Clone: JDC-14
Isotype: IgG1, κ
Species Reactivity: Human
ELISA
Specifications
Specifications
| Antigen | Mouse anti-Human IgG4 |
| Classification | Monoclonal |
| Clone | JDC-14 |
| Conjugate | Alkaline Phosphatase (AP) |
| Host Species | Mouse |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Target Species | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?